Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAST1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584915
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584915 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584915 Supplier Invitrogen™ Supplier No. PA584915
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LTLREKTWRG GSPEIKRFSA SEASFLEGEA SPPLGARRRF SALLEPSRFS APQEDEDE Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.

Appears to link the dystrophin/utrophin network with microtubule filaments via the syntrophins. Phosphorylation of DMD or UTRN may modulate their affinities for associated proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAST1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAST1
Gene Accession No. Q9Y2H9
Gene Alias 9430008B02Rik; KIAA0973; MAST1; mast1 {ECO:0000312; microtubule associated serine/threonine kinase 1; microtubule-associated serine/threonine-protein kinase 1; RGD:631372}; SAST; SAST170; syntrophin associated serine/threonine kinase; syntrophin-associated serine/threonine-protein kinase; syntrophin-associated serine-threonine protein kinase
Gene Symbols MAST1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAST1. Recombinant protein control fragment (Product #RP-108262).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22983
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.