Learn More
Invitrogen™ MAST1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114244
Description
Immunogen sequence: QESPLSLGAD PLLPEGASRP PVSSKEKESP GGAEACTPPR ATTPGGRTLE For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 93%; Human: 100%; Pig: 100%.
Appears to link the dystrophin/utrophin network with microtubule filaments via the syntrophins. Phosphorylation of DMD or UTRN may modulate their affinities for associated proteins.
Specifications
| MAST1 | |
| Polyclonal | |
| Unconjugated | |
| MAST1 | |
| 9430008B02Rik; KIAA0973; MAST1; mast1 {ECO:0000312; microtubule associated serine/threonine kinase 1; microtubule-associated serine/threonine-protein kinase 1; RGD:631372}; SAST; SAST170; syntrophin associated serine/threonine kinase; syntrophin-associated serine/threonine-protein kinase; syntrophin-associated serine-threonine protein kinase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22983 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y2H9 | |
| MAST1 | |
| Synthetic peptide directed towards the C-terminal region of Human MAST1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.