Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAST2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501561995
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-1995 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-1995 Supplier Bethyl Laboratories, Inc Supplier No. A302188A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Syntrophin is an adapter protein that functions to bind certain signaling molecules to the dystrophin-associated protein complex. This complex connects the extracellular matrix to the intracellular cytoskeleton for construction and maintece of the postsynaptic structures in the neuromuscular junction and the central nervous system. Microtubule-associated serine/threonine-protein kinase 2 (MAST205) is a testis-specific, cytoplasmic protein that functions in a multi-protein complex in the maturation of spermatids. MAST205 is involved in linking the dystrophin/utrophin network with microtubule filaments via Syntrophin. By forming a complex with TRAF6, MAST205 regulates lipopolysaccharide-induced IL-12 synthesis in macrophages. This leads to the inhibition of TRAF6 NF-kappa-B activation. Two isoforms exist for MAST205 due to alternative splicing. Isoform 1 represents the full length protein, while isoform 2 lacks the residues 327-396 and 1091-1113. The N-terminus of MAST205 must be phosphorylated in order for ubiquitination to occur at the same site. This ubiquitination leads to the degradation of MAST205 via proteasome-mediated proteolysis.

Specifications

Antigen MAST2
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene MAST2
Gene Accession No. Q6P0Q8
Gene Alias KIAA0807, MAST205
Gene Symbols MAST2
Host Species Rabbit
Immunogen Between 25 and 75
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23139
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.