Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAST205 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP122986AF405
Description
MAST205 Polyclonal antibody specifically detects MAST205 in Human samples. It is validated for Western BlotSpecifications
MAST205 | |
Polyclonal | |
Western Blot | |
EC 2.7.11, EC 2.7.11.1, FLJ39200, KIAA0807RP4-533D7.1, MAST205microtubule associated testis specific serine/threonine protein kinase, microtubule associated serine/threonine kinase 2, microtubule-associated serine/threonine-protein kinase 2, MTSSK | |
The immunogen recognized by this antibody maps to a region between residue 1748 and 1798 of human microtubule associated serine/threonine kinase 2 using the numbering given in entry NP_055927.2 (GeneID 23139). | |
0.1 mL | |
Protein Kinase | |
23139 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction