Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAST3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599246
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599246 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599246 Supplier Invitrogen™ Supplier No. PA599246
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MAST3.

MAST3 gene ontology annotations related to this gene include cytoskeleton organization; intracellular signal transduction; peptidyl-serine phosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAST3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MAST3
Gene Accession No. O60307, Q3U214
Gene Alias BC024265; Kiaa0561; MAST3; microtubule associated serine/threonine kinase 3; microtubule-associated serine/threonine-protein kinase 3; mKIAA0561
Gene Symbols MAST3
Host Species Rabbit
Immunogen A synthesized peptide derived from human MAST3(Accession O60307), corresponding to amino acid residues G1045-G1095.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23031, 546071
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.