Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAST3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536975
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536975 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536975 Supplier Invitrogen™ Supplier No. PA536975
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 143 kDa. Purity is >95% by SDS-PAGE.

MAST3 gene ontology annotations related to this gene include cytoskeleton organization; intracellular signal transduction; peptidyl-serine phosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAST3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene MAST3
Gene Accession No. O60307, Q3U214
Gene Alias BC024265; Kiaa0561; MAST3; microtubule associated serine/threonine kinase 3; microtubule-associated serine/threonine-protein kinase 3; mKIAA0561
Gene Symbols MAST3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 1075-1120 of human MAST3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23031, 546071, 688540
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.