Learn More
Eppendorf™ Masterclear™ Real-time PCR Film PROMO
Catalog No. 14285820P Shop All Eppendorf North America Products
Change view
Click to view available options
Eppendorf™ Real-time PCR Film
Catalog No.
14-285-820P Eppendorf™ Real-time PCR Film
1 options
Catalog No. 14-285-820P Supplier Eppendorf™ Supplier No. 2231001132
Optimized for maximum light transmission and tight sealing
  • Peelable design
  • PCR clean purity grade
TRUSTED_SUSTAINABILITY
Specifications
Product Type Real-time PCR Film