Learn More
StatLab™ MasterTech Alcian Blue PAS Stain Kit
SDP
Catalog No. STLKTAPA
Change view
Click to view available options
Quantity:
100 mL
1 pt.
Catalog No. Quantity
STLKTAPA 100 mL
STLKTAPAPT 1 pt.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLKTAPA
Supplier: StatLab™ KTAPA
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
PAS Stain Kit stains acid mucins, neutral mucins, and glycogen in 60 min.
  • Alcian Blue and Schiff's serve as the primary stains, producing blue to purple stained acid mucins and pink to red neutral mucins or glycogen with blue nuclei counterstained by Hematoxylin
  • 100 mL Stain Kit yields between 80 to 100 slides
  • 1 pt. Stain Kit yields between 320 to 500 slides
Specifications
Type Stain Kit
Quantity 100 mL