Learn More
StatLab™ MasterTech Alcian Blue Stain Kit
SDP
Catalog No. p-200020485
Click to view available options
Quantity:
100 mL
1 pt.
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Alcian Blue Stain Kit stains acid mucins blue in 40 minutes.
  • With a 2.5 pH, this kit contains a specially formulated Nuclear Fast Red that reduces flocculation, and greatly reduces staining artifacts and debris
  • Alcian Blue serves as the primary stain, producing vibrant blue stained mucins with the pink to red nuclei counterstained by Nuclear Fast Red
  • 100 mL Stain Kit yields between 80 and 100 slides
  • 1 pt. Stain Kit yields between 320 and 500 slides
Specifications
Type Stain Kit
Quantity 100 mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.