Learn More
StatLab™ MasterTech GMS Stain Kit
Click to view available options
Quantity:
100 mL
1 pt.
Description
- Kit includes a 15 minute microwave procedure and the traditional one hour and thirty minute procedure
- A Methenamine Silver Developer Solution serves as the primary stain, with Gold Chloride toning the tissue sections, producing gray to black fungi and Pneumocystis jirovecii against a green background counterstained with Light Green
- 100 mL Stain Kit yields between 20 and 32 slides
- 1 pt. Stain Kit yields between 100 and 400 slides
Specifications
Specifications
Type | Stain Kit |
Quantity | 1 pt. |