Learn More
StatLab™ MasterTech Nuclear Fast Red Stain
Click to view available options
Quantity:
1 L
1 gal./Bottle, 4 Bottles/Cs.
1 pt.
100 mL
5 gal. Bottle
Description
- For the staining of the nuclei where it will react with negatively charged components such as nucleic acids in the nucleus
- An ideal alternative to hematoxylin which would otherwise obscure the substrate colors
Specifications
Specifications
Type | Nuclear Stain |
Quantity | 1 pt. |