Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
StatLab MasterTech Special Stain Kit, Gold Chloride Powder

Click to view available options
:
1 g
Description
- Used in histology for staining and visualization of neurons and nerve fibers
- It acts as a reducing agent in various silver plating methods
Specifications
Specifications
| Physical Form | Aqueous Solution |
| Color | Golden/Green |
| Quantity | 1 g |
| Grade | ACS |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction