Learn More
StatLab™ MasterTech Trichrome Masson's Stain Kit
Click to view available options
Quantity:
100 mL
1 pt.
Description
- Biebrich Scarlet/Acid Fuchsin and Aniline Blue serve as the primary stains, producing red cytoplasm, intercellular fiber, keratin, and muscle, blue collagen and mucus with purple to black nuclei; counterstained by Weigert's Hematoxylin
- 100 mL Stain Kit yields between 80 and 100 slides
- 1 pt. Stain Kit yields between 320 and 500 slides
Specifications
Specifications
Type | Stain Kit |
Quantity | 1 pt. |