Learn More
StatLab™ MasterTech Trichrome Masson's Stain Kit
SDP
Catalog No. p-200020497
Click to view available options
Quantity:
100 mL
1 pt.
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Masson's Stain Kit stains for collagen and muscle in as little 20 minutes.
  • Biebrich Scarlet/Acid Fuchsin and Aniline Blue serve as the primary stains, producing red cytoplasm, intercellular fiber, keratin, and muscle, blue collagen and mucus with purple to black nuclei; counterstained by Weigert's Hematoxylin
  • 100 mL Stain Kit yields between 80 and 100 slides
  • 1 pt. Stain Kit yields between 320 and 500 slides
Specifications
Type Stain Kit
Quantity 100 mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.