Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAT1A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $646.00
Specifications
Antigen | MAT1A |
---|---|
Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
MAT1A Polyclonal specifically detects MAT1A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MAT1A | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
Q00266 | |
4143 | |
This antibody was developed against a recombinant protein corresponding to amino acids: MNGPVDGLCDHSLSEGVFMFTSESV | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Lipid and Metabolism | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
AdoMet synthase 1, adoMet synthetase 1, AMS1, MAT, MATA1MAT 1, MAT-I/III, Methionine adenosyltransferase 1, methionine adenosyltransferase I, alpha, Methionine adenosyltransferase I/III, S-adenosylmethionine synthase isoform type-1, S-adenosylmethionine synthetase isoform type-1, SAMS, SAMS1EC 2.5.1.6 | |
MAT1A | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title