Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAT1A/MAT2A Recombinant Rabbit Monoclonal Antibody (HL1679)
GREENER_CHOICE

Catalog No. PIMA547139
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547139 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547139 Supplier Invitrogen™ Supplier No. MA547139
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

S adenosylmethionine synthetase catalyzes the formation of S adenosylmethionine from methionine and ATP. In mammalian tissues, there are three distinct forms of AdoMet synthases designated as alpha, beta, and gamma. Alpha and beta are expressed only in adult liver, while gamma is widely distributed in extrahepatic tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAT1A/MAT2A
Applications Western Blot
Classification Recombinant Monoclonal
Clone HL1679
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Mat2a
Gene Accession No. P31153, Q00266, Q3THS6, Q91X83
Gene Alias AdoMet; adoMet synthase 1; AdoMet synthase 2; adoMet synthetase 1; adoMet synthetase 2; AI046368; Ams; Ams1; Ams2; D630045P18Rik; MAT; MAT 1; MAT 2; Mat1a; Mat2a; MATA1; MATA2; MAT-I/III; MATII; MAT-II; Methionine adenosyltransferase 1; methionine adenosyltransferase 1A; methionine adenosyltransferase 2; methionine adenosyltransferase 2A; methionine adenosyltransferase I, alpha; methionine adenosyltransferase I/III; Methionine adenosyltransferase II; methionine adenosyltransferase II, alpha; non-hepatic-type S-adenosylmethionine synthetase; S - adenosylmethionine synthetase; SADE; S-adenosylmethionine synthase isoform type-1; S-adenosylmethionine synthase isoform type-2; S-adenosylmethionine synthetase isoform type-1; S-adenosylmethionine synthetase isoform type-2; SAMS; SAMS1; Sams2; SAS; testicular tissue protein Li 121
Gene Symbols Mat1a, Mat2a
Host Species Rabbit
Immunogen Recombinant fragment of human MAT1A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11720, 232087, 4143, 4144
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.