Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAT1A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541998
Description
Peptide sequence: TSESVGEGHP DKICDQISDA VLDAHLKQDP NAKVACETVC KTGMVLLCGE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
MAT1A catalyzes the formation of S-adenosylmethionine from methionine and ATP. Methionine adenosyltransferase deficiency is caused by recessive and dominant mutations, the latter identified in autosomal dominant persistant hypermethioninemia.This gne encodes methionine adenosyltransferase I (alpha isoform), which catalyzes the formation of S-adenosylmethionine from methionine and ATP. Methionine adenosyltransferase deficiency is caused by recessive and dominant mutations, the latter identified in autosomal dominant persistant hypermethioninemia.
Specifications
MAT1A | |
Polyclonal | |
Unconjugated | |
MAT1A | |
AdoMet; AdoMet synthase 1; adoMet synthetase 1; AI046368; Ams; Ams1; Cartilage matrix protein; CMP; CRTM; MAT; MAT 1; Mat1a; MATA1; MAT-I/III; MATN1; matrilin 1, cartilage matrix protein; Matrilin-1; Methionine adenosyltransferase 1; methionine adenosyltransferase 1A; methionine adenosyltransferase I, alpha; methionine adenosyltransferase I/III; S - adenosylmethionine synthetase; SADE; S-adenosylmethionine synthase; S-adenosylmethionine synthase isoform type-1; S-adenosylmethionine synthase isoform type-1; S-adenosylmethionine synthase; S-adenosylmethionine synthetase isoform type-1; SAMS; SAMS1; SAS; similar to S-adenosylmethionine synthetase alpha and beta forms (Methionine adenosyltransferase) (AdoMet synthetase) (MAT-I/III); wu:fi35e01; zgc:55442 | |
Rabbit | |
Protein A | |
RUO | |
4143 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q00266 | |
MAT1A | |
Synthetic peptide directed towards the N-terminal of human MAT1A (aa 21-70). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction