Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAT1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116973
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116973 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116973 Supplier Invitrogen™ Supplier No. PA5116973
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse liver, Rat liver Immunogen sequence: MNGPVDGLCD HSLSEGVFMF TSESVGEGHP DKICDQISDA VLDAHLKQDP NAKVACETVC KTGMVLLCGE ITSMAMVDYQ RVVRDTIKHI GYDDSAKGFD FKTCNVLVAL EQQSPDIAQC VHLDRNEEDV GAGDQGLMFG YATDETEECM PLTIILAHKL NARMADLRRS GLLPWLRPDS KTQVTVQYMQ DNGAVIPVRI HTIVISVQHN EDITLEEMRR ALKEQVIRAV VPAKYLDEDT VYHLQPSGRF VIGGPQGDAG VTGRKIIVDT YGGWGAHGGG AFSGKDYTKV DRSAAYAARW VAKSLVKAGL CRRVLVQVSY AIGVAEPLSI SIFTYGTSQK TERELLDVVH KNFDLRPGVI VRDLDLKKPI YQKTACYGHF GRSEFPWEVP RKLVF.

MAT1A catalyzes the formation of S-adenosylmethionine from methionine and ATP. Methionine adenosyltransferase deficiency is caused by recessive and dominant mutations, the latter identified in autosomal dominant persistant hypermethioninemia.This gne encodes methionine adenosyltransferase I (alpha isoform), which catalyzes the formation of S-adenosylmethionine from methionine and ATP. Methionine adenosyltransferase deficiency is caused by recessive and dominant mutations, the latter identified in autosomal dominant persistant hypermethioninemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAT1A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MAT1A
Gene Accession No. P13444, Q00266, Q91X83
Gene Alias AdoMet; AdoMet synthase 1; adoMet synthetase 1; AI046368; Ams; Ams1; Cartilage matrix protein; CMP; CRTM; MAT; MAT 1; Mat1a; MATA1; MAT-I/III; MATN1; matrilin 1, cartilage matrix protein; Matrilin-1; Methionine adenosyltransferase 1; methionine adenosyltransferase 1A; methionine adenosyltransferase I, alpha; methionine adenosyltransferase I/III; S - adenosylmethionine synthetase; SADE; S-adenosylmethionine synthase; S-adenosylmethionine synthase isoform type-1; S-adenosylmethionine synthase isoform type-1; S-adenosylmethionine synthase; S-adenosylmethionine synthetase isoform type-1; SAMS; SAMS1; SAS; similar to S-adenosylmethionine synthetase alpha and beta forms (Methionine adenosyltransferase) (AdoMet synthetase) (MAT-I/III); wu:fi35e01; zgc:55442
Gene Symbols MAT1A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-395 of human MAT1A (NP_0004201).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11720, 25331, 4143
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.