Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAT1A Rabbit anti-Human, Mouse, Plant, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501727954 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7954 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7954 Supplier Proteintech Group Inc Supplier No. 123951AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody was raised against the full length of human MAT1A and may cross-react with MAT2A. Western blot analysis using this antibody detected a single band around 44-50kD in liver.

MAT1A catalyzes the formation of S-adenosylmethionine from methionine and ATP. Methionine adenosyltransferase deficiency is caused by recessive and domit mutations, the latter identified in autosomal domit persistant hypermethioninemia.This gne encodes methionine adenosyltransferase I (alpha isoform), which catalyzes the formation of S-adenosylmethionine from methionine and ATP. Methionine adenosyltransferase deficiency is caused by recessive and domit mutations, the latter identified in autosomal domit persistant hypermethioninemia.

Specifications

Antigen MAT1A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MAT1A
Gene Accession No. P13444, Q00266, Q91X83
Gene Alias AdoMet synthase 1, AMS1, MAT, MAT 1, MAT I/III, MAT1A, MATA1, SAMS, SAMS1
Gene Symbols MAT1A
Host Species Rabbit
Immunogen MAT1A Fusion Protein Ag3076
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11720, 25331, 4143
Target Species Human, Mouse, Plant, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.