Learn More
Description
Specifications
Specifications
| Antigen | MAT2A |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml, Simple Western, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:2000, Immunoprecipitation reported in scientific literature (PMID 25294683), Immunohistochemistry-Paraffin 1:200 |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | P31153 |
| Gene Alias | AdoMet synthase 2, AMS2, EC 2.5.1.6, MAT 2, MATA2SAMS2adoMet synthetase 2, MATII, MAT-II, Methionine adenosyltransferase 2, Methionine adenosyltransferase II, methionine adenosyltransferase II, alpha, S-adenosylmethionine synthase isoform type-2, S-adenosylmethionine synthetase isoform type-2 |
| Gene Symbols | MAT2A |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
