Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAT2A Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11094158H
Description
MAT2A Polyclonal specifically detects MAT2A in Human, Mouse, Rat, Porcine, Bovine, Primate samples. It is validated for Western Blot, Simple Western.Specifications
MAT2A | |
Polyclonal | |
Western Blot, Simple Western | |
P31153 | |
MAT2A | |
Synthetic peptide made to an N-terminal portion of human MAT2A (within residues 1-100). [Swiss-Prot# P31153] | |
Affinity Purified | |
RUO | |
Primary | |
Human, rat, mouse, orangutan, bovine and monkey. Immunogen sequence has 94% homology to porcine and chicken and 86% homology to Zebrafish. | |
Store at 4C in the dark. |
Western Blot | |
HRP | |
PBS with No Preservative | |
AdoMet synthase 2, AMS2, EC 2.5.1.6, MAT 2, MATA2SAMS2adoMet synthetase 2, MATII, MAT-II, Methionine adenosyltransferase 2, Methionine adenosyltransferase II, methionine adenosyltransferase II, alpha, S-adenosylmethionine synthase isoform type-2, S-adenosylmethionine synthetase isoform type-2 | |
Rabbit | |
43 kDa | |
0.1 mL | |
Amino Acids Drugs and other small molecules, Cancer, Lipid and Metabolism, Signal Transduction | |
4144 | |
Human, Mouse, Rat, Pig, Bovine, Primate | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction