Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAT2A Antibody (SR1837), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | MAT2A |
---|---|
Clone | SR1837 |
Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Description
MAT2A Monoclonal antibody specifically detects MAT2A in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
MAT2A | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
AdoMet synthase 2, AMS2, EC 2.5.1.6, MAT 2, MATA2SAMS2adoMet synthetase 2, MATII, MAT-II, Methionine adenosyltransferase 2, Methionine adenosyltransferase II, methionine adenosyltransferase II, alpha, S-adenosylmethionine synthase isoform type-2, S-adenosylmethionine synthetase isoform type-2 | |
A synthesized peptide derived from human MAT2A (Uniprot #: P31153) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1837 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Amino Acids Drugs and other small molecules, Cancer, Lipid and Metabolism, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
4144 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title