Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAT2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115550
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115550 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115550 Supplier Invitrogen™ Supplier No. PA5115550
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MAT2A.

The protein encoded by this gene catalyzes the production of S-adenosylmethionine (AdoMet) from methionine and ATP. AdoMet is the key methyl donor in cellular processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAT2A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene MAT2A
Gene Accession No. P18298, P31153, Q3THS6
Gene Alias adoMet synthase 2; adoMet synthetase 2; AMS2; D630045P18Rik; MAT 2; MAT2A; MATA2; MATII; MAT-II; Methionine adenosyltransferase 2; methionine adenosyltransferase 2A; Methionine adenosyltransferase II; methionine adenosyltransferase II, alpha; non-hepatic-type S-adenosylmethionine synthetase; S-adenosylmethionine synthase; S-adenosylmethionine synthase isoform type-2; S-adenosylmethionine synthetase isoform type-2; Sams2; testicular tissue protein Li 121
Gene Symbols MAT2A
Host Species Rabbit
Immunogen A synthesized peptide derived from human MAT2A(Accession P31153), corresponding to amino acid residues P357-Y395.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171347, 232087, 4144
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.