Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAT2A Polyclonal Antibody

Catalog No. PIPA5121780
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121780 20 μL
PIPA5119281 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121780 Supplier Invitrogen™ Supplier No. PA5121780
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MAT2A Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

The protein encoded by this gene catalyzes the production of S-adenosylmethionine (AdoMet) from methionine and ATP. AdoMet is the key methyl donor in cellular processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAT2A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene MAT2A
Gene Accession No. P31153
Gene Alias adoMet synthase 2; adoMet synthetase 2; AMS2; D630045P18Rik; MAT 2; MAT2A; MATA2; MATII; MAT-II; Methionine adenosyltransferase 2; methionine adenosyltransferase 2A; Methionine adenosyltransferase II; methionine adenosyltransferase II, alpha; non-hepatic-type S-adenosylmethionine synthetase; S-adenosylmethionine synthase; S-adenosylmethionine synthase isoform type-2; S-adenosylmethionine synthetase isoform type-2; Sams2; testicular tissue protein Li 121
Gene Symbols MAT2A
Host Species Rabbit
Immunogen Recombinant protein Methionine Adenosyltransferase II Alpha. The antigen corresponds to amino acid range 176-395 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4144
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.