Learn More
MAT2A Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304279AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat and Orangutan
Catalyzes the formation of S-adenosylmethionine from methionine and ATP.Specifications
MAT2A | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P31153, Q3THS6 | |
Mat2a | |
Between 1 and 50 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
MAT2A | |
MATA2, MATII, SAMS2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
232087, 4144 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.