Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAT2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558075
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558075 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558075 Supplier Invitrogen™ Supplier No. PA558075
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MFDKVQFSNK SANMDHWQQR FPTHVKDVAT VCRQLAEKRM LDPSIKGTFH WSGNEQMTKY EMACAIADAF N Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

MAT2B belongs to the methionine adenosyltransferase (MAT) family. MAT catalyzes the biosynthesis of S-adenosylmethionine from methionine and ATP. This protein is the regulatory beta subunit of MAT. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAT2B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAT2B
Gene Accession No. Q9NZL9
Gene Alias 1110064C04Rik; 2410018D16Rik; AI182287; AU022853; beta regulatory subunit of methionine adenosyltransferase; dTDP-4-keto-6-deoxy-D-glucose 4-reductase; MAT II beta; MAT2B; MAT-II; MATIIbeta; methionine adenosyltransferase 2 beta subunit; methionine adenosyltransferase 2 subunit beta; methionine adenosyltransferase 2B; Methionine adenosyltransferase II beta; methionine adenosyltransferase II, beta; MSTP045; Nbla02999; Putative dTDP-4-keto-6-deoxy-D-glucose 4-reductase; putative protein product of Nbla02999; SDR23E1; short chain dehydrogenase/reductase family 23E, member 1; testicular tissue protein Li 118; TGR; UNQ2435/PRO4995
Gene Symbols MAT2B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAT2B. Recombinant protein control fragment (Product #RP-96793).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27430
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.