Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MATE 2 Rabbit anti-Canine, Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 268731AP150UL

Description
S47A2 belongs to a family of transporters involved in excretion of toxic electrolytes, both endogenous and exogenous, through urine and bile. This transporter family shares homology with the bacterial MATE (multidrug and toxin extrusion) protein family responsible for drug resistance. This gene is one of two members of the MATE transporter family located near each other on chromosome 17.Specifications
MATE 2 | |
Polyclonal | |
Unconjugated | |
SLC47A2 | |
hMATE 2, MATE 2, MATE2, MATE2 B, MATE2 K, SLC47A2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
146802, 489527 | |
-20°C | |
Liquid |
Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q86VL8 | |
SLC47A2 | |
MATE 2 Fusion Protein Ag25472 | |
150 μL | |
Primary | |
Canine, Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction