Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Math Marks the Spot™ Math Activity Game
SCI_ED

Catalog No. s90833
Click to view available options
:
Math Marks the Spot Game

Catalog No. S90833

Supplier: Learning Resources, Inc LER0383

Only null left
Add to Cart
Add to Cart

Includes

3 cubes and 28 cardboard game markers.

Shake, roll and race!

Roll the soft foam numbers and operation cubes and race to find the answer on the oversized number mat!

  • Features a durable, vinyl mat
  • Displays all the answers (sums and differences) to addition and subtraction problems that use numbers 1 through 6

Order Info

Grades K+.

Specifications

Grade Level Elementary
Grade K
For Use With (Application) Practice addition and subtraction problems that use numbers 1 through 6
Includes Durable, vinyl number mat; Foam numbers and operation cubes; 28 cardboard game markers
Description Math Marks the Spot™ Math Activity Game
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.