Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Math Mat Challenge™ Game
SCI_ED

Catalog No. s79392
Change view
Click to view available options
:
Math
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S79392 Math
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S79392 Supplier Learning Resources, Inc Supplier No. LER 0047

Requires

Three AA batteries

Interactive talking floor mat with quiz games makes learning math fun.

Listen to the equation, do the math, then step on the right answer.

  • Six different games
  • Two skill levels
  • Reinforces numbers, counting, addition and subtraction up to the number 20.
  • Dimensions: 34 in. dia. (85cm)

Order Info

Grades PK-2

Specifications

Description Math Mat Challenge™ Game
Grade Pre-K to 2
For Use With (Application) Reinforces numbers, counting, addition and subtraction up to the number 20
Includes Six different games, Two skill levels
Size 34 in. (85cm) dia.
Diameter (English) Outer 34 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.