Learn More
Invitrogen™ ATOH1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593035
Description
Immunogen sequence: LQTPSGGEQP PPPPASCKSD HHHLRTAASY EGGAGNATAA GAQQASGGSQ RPTPPGSCRT RFSAPASAGG YSVQLDALHF STFEDSALTA MMAQKNLSPS LPGSILQPVQ EENSKTSPRS HRSDGEFSPH SHYSDSDEAS; Positive Samples: SW620, HepG2, human pacenta, Mouse liver, Mouse heart; Cellular Location: Nucleus.
The Drosophila atonal gene produces a protein with basic helix loop helix (bHLH) domains that plays an essential role in the development of the Drosophila nervous system. Mammalian atonal homolog 1 (MATH-1) is a helix-loop-helix (HLH) transcription factor that is structurally homologous to the product of the Drosophila proneural gene atonal. MATH-1, so known as Atoh1, Ath1 or HATH-1, is a 351 amino acid protein with an atonal-related basic HLH domain. In mice, expression of MATH-1 takes place by embryonic day 9. 5 and initially localizes to the cranial ganglions and the dorsal part of the central nervous system. Prominent expression of MATH-1 is in the dorsal part of the central nervous system but becomes restricted to the external granular layer of the cerebellum by day 18 and is undetectable in the adult nervous system. It is suggested that MATH-1 may play a role in the differentiation of subsets of neural cells by activating E box-dependent transcription.
Specifications
ATOH1 | |
Polyclonal | |
Unconjugated | |
Atoh1 | |
ATH1; Atoh1; atonal bHLH transcription factor 1; atonal homolog 1; atonal homolog 1 (Drosophila); atonal homolog bHLH transcription factor 1; BHLHA14; Class A basic helix-loop-helix protein 14; H MATH-1; Hath1; Helix-loop-helix protein hATH-1; helix-loop-helix protein mATH-1; Math1; MATH-1; Protein atonal homolog 1; RGD1565171 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
11921, 474 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.84 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P48985, Q92858 | |
Atoh1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 215-354 of human ATOH1 (NP_0051631). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.