Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATOH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597480
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597480 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597480 Supplier Thermo Scientific Supplier No. PA597480
Add to Cart
Add to Cart

MATH1 Polyclonal Antibody

MATH1 Polyclonal Antibody for Western Blot, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen ATOH1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Atoh1
Gene Accession No. P48985
Gene Alias ATH1; Atoh1; atonal bHLH transcription factor 1; atonal homolog 1; atonal homolog 1 (Drosophila); atonal homolog bHLH transcription factor 1; BHLHA14; Class A basic helix-loop-helix protein 14; H MATH-1; Hath1; Helix-loop-helix protein hATH-1; helix-loop-helix protein mATH-1; Math1; MATH-1; Protein atonal homolog 1; RGD1565171
Gene Symbols Atoh1
Host Species Rabbit
Immunogen Recombinant Mouse Protein atonal homolog 1 protein (1-351AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11921
Target Species Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.