Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATOH1 Recombinant Rabbit Monoclonal Antibody (JE60-25)
GREENER_CHOICE

Catalog No. PIMA544829
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544829 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544829 Supplier Invitrogen™ Supplier No. MA544829
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

ATOH1 Recombinant Monoclonal Antibody for Western Blot

The Drosophila atonal gene produces a protein with basic helix loop helix (bHLH) domains that plays an essential role in the development of the Drosophila nervous system. Mammalian atonal homolog 1 (MATH-1) is a helix-loop-helix (HLH) transcription factor that is structurally homologous to the product of the Drosophila proneural gene atonal. MATH-1, so known as Atoh1, Ath1 or HATH-1, is a 351 amino acid protein with an atonal-related basic HLH domain. In mice, expression of MATH-1 takes place by embryonic day 9. 5 and initially localizes to the cranial ganglions and the dorsal part of the central nervous system. Prominent expression of MATH-1 is in the dorsal part of the central nervous system but becomes restricted to the external granular layer of the cerebellum by day 18 and is undetectable in the adult nervous system. It is suggested that MATH-1 may play a role in the differentiation of subsets of neural cells by activating E box-dependent transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATOH1
Applications Western Blot
Classification Recombinant Monoclonal
Clone JE60-25
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Atoh1
Gene Accession No. P48985, Q92858
Gene Alias ATH1; Atoh1; atonal bHLH transcription factor 1; atonal homolog 1; atonal homolog 1 (Drosophila); atonal homolog bHLH transcription factor 1; BHLHA14; Class A basic helix-loop-helix protein 14; H MATH-1; Hath1; Helix-loop-helix protein hATH-1; helix-loop-helix protein mATH-1; Math1; MATH-1; Protein atonal homolog 1; RGD1565171
Gene Symbols Atoh1
Host Species Rabbit
Immunogen Synthetic peptide within human ATOH1 aa 1-50.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11921, 474, 500156
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.