Learn More
Invitrogen™ MATK Polyclonal Antibody

Description
Immunogen sequence: GTQCITKCEH TRPKPGELAF RKGDVVTILE ACENKSWYRV KHHTSGQEGL LAAGALRERE ALSADPKLSL MPWFHGKISG QEAVQQLQPP EDGLFLVRES ARHPGDYVLC VSFGRDVIHY RVLHRDGHLT IDEAVFFCNL MDMVEHY Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 92%.
Specifications
Specifications
| Antigen | MATK |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MATK |
| Gene Accession No. | P42679 |
| Gene Alias | Batk; CHK; Csk homologous kinase; Csk-homologous kinase; Csk-type protein tyrosine kinase; CTK; DKFZp434N1212; Hematopoietic consensus tyrosine-lacking kinase; HHYLTK; hydroxyaryl-protein kinase; HYL; HYL tyrosine kinase; HYLTK; leukocyte carboxyl-terminal src kinase related; Lsk; Matk; megakaryocyte-associated tyrosine kinase; megakaryocyte-associated tyrosine kinase (non-receptor protein tyrosine kinase); megakaryocyte-associated tyrosine-protein kinase; MGC1708; MGC2101; non-receptor protein kinase protein; Ntk; Protein kinase BATK; protein kinase HYL; Protein kinase NTK; tyrosine kinase MATK; tyrosine-protein kinase CTK; tyrosylprotein kinase |
| Gene Symbols | MATK |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.