Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MATK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550179
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550179 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550179 Supplier Invitrogen™ Supplier No. PA550179
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total MATK protein.

MATK (Hyl) is a non-receptor CSK-type protein tyrosine kinase that is thought to function in signal transduction pathways important in megakaryocyte growth and/or differentiation. This protein is thought to play a significant role in the signal transduction of hematopoietic cells. It is able to phosphorylate and inactivate Src family kinases, and may play an inhibitory role in the control of T-cell proliferation. This protein might be involved in signaling in some cases of breast cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MATK
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MATK
Gene Accession No. P41242, P41243, P42679
Gene Alias Batk; CHK; Csk homologous kinase; Csk-homologous kinase; Csk-type protein tyrosine kinase; CTK; DKFZp434N1212; Hematopoietic consensus tyrosine-lacking kinase; HHYLTK; hydroxyaryl-protein kinase; HYL; HYL tyrosine kinase; HYLTK; leukocyte carboxyl-terminal src kinase related; Lsk; Matk; megakaryocyte-associated tyrosine kinase; megakaryocyte-associated tyrosine kinase (non-receptor protein tyrosine kinase); megakaryocyte-associated tyrosine-protein kinase; MGC1708; MGC2101; non-receptor protein kinase protein; Ntk; Protein kinase BATK; protein kinase HYL; Protein kinase NTK; tyrosine kinase MATK; tyrosine-protein kinase CTK; tyrosylprotein kinase
Gene Symbols MATK
Host Species Rabbit
Immunogen Synthesized peptide derived from Internal of human MATK.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17179, 4145, 60450
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.