Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MATK Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548676
Description
Predicted to react with rat based on sequence homology.
MATK has amino acid sequence similarity to Csk tyrosine kinase and has the structural features of the CSK subfamily: SRC homology SH2 and SH3 domains, a catalytic domain, a unique N terminus, lack of myristylation signals, lack of a negative regulatory phosphorylation site, and lack of an autophosphorylation site. This protein is thought to play a significant role in the signal transduction of hematopoietic cells. It is able to phosphorylate and inactivate Src family kinases, and may play an inhibitory role in the control of T-cell proliferation. This protein might be involved in signaling in some cases of breast cancer.
Specifications
| Matk | |
| Polyclonal | |
| PBS with 0.09% sodium azide | |
| P41242 | |
| MATK | |
| KLH conjugated synthetic peptide between 477-505 amino acids from the C-terminal region of mouse Matk. | |
| 400 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Matk | |
| Megakaryocyte-associated tyrosine-protein kinase, Protein kinase NTK, Tyrosine-protein kinase CTK, Matk, Ctk, Ntk | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17179 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction