Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MATK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 100822AP150UL

Description
All members of the Src gene family of tyrosine kinases are characterized by a carboxy terminal domain tyrosine, Y527 in the case of Src p60, which is highly phosphorylated in the inactive form of the enzyme, while phosphorylated to a much lesser extent when the enzyme is active. For instance, a mutant of c-Src, in which Y527 is replaced by phenylalanine, is transforming and displays 5 to 10-fold elevated kinase activity compared to its normal counterpart. Csk has been identified as a Src related tyrosine kinase having both SH2 and SH3 domains and a catalytic domain but lacking sequences amino terminal to the SH3 domain as well as the carboxy terminal regulatory sequences. Csk phosphorylates Src on Y527 and also down regulates Fyn, Fyn and Lck by tyrosine phosphorylation of carboxy terminal regulatory sites. A Csk-like protein-tyrosine kinase of mouse cell origin (Ctk), also designated Ntk, and its human homolog, Lsk, have also been described.Specifications
MATK | |
Polyclonal | |
Unconjugated | |
MATK | |
CHK, CTK, HHYLTK, HYL, HYLTK, Lsk, MATK, Protein kinase HYL, Tyrosine protein kinase CTK | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17179, 4145, 60450 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P41242, P41243, P42679 | |
Matk | |
MATK Fusion Protein Ag0128 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction