Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MATN1 Monoclonal Antibody (5A8)
GREENER_CHOICE

Catalog No. PIMA515704
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515704 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515704 Supplier Invitrogen™ Supplier No. MA515704
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15704 targets MATN1 in indirect ELISA, IF and WB applications and shows reactivity with Human samples. The MA5-15704 immunogen is purified recombinant fragment of human MATN1 expressed in E. Coli. MA5-15704 detects MATN1 which has a predicted molecular weight of approximately 54kDa.

This gene catalyzes a two-step reaction that involves the transfer of the adenosyl moiety of ATP to methionine to form S-adenosylmethionine and tripolyphosphate, which is subsequently cleaved to PPi and Pi. S-adenosylmethionine is the source of methyl groups for most biological methylations. The encoded protein is found as a homotetramer or a homodimer whereas a third form, MAT II , is encoded by the MAT2A gene. Mutations in this gene are associated with methionine adenosyltransferase deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MATN1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 5A8
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene MATN1
Gene Accession No. P21941
Gene Alias cartilage matrix protein; Cmp; CRTM; MAT; Mat1; MAT1A; MATA1; MATN1; matrilin 1, cartilage matrix protein; matrilin 1, cartilage matrix protein 1; matrilin-1; SAMS; SAMS1
Gene Symbols MATN1
Host Species Mouse
Immunogen Purified recombinant fragment of human MATN1 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4146
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.