Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MATN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583020
Description
Immunogen sequence: REIASEPVAE HYFYTADFKT INQIGKKLQK KICVEEDPCA CESLVKFQAK VEGLLQALTR KLEAVSKRLA ILENTVV.
This gene catalyzes a two-step reaction that involves the transfer of the adenosyl moiety of ATP to methionine to form S-adenosylmethionine and tripolyphosphate, which is subsequently cleaved to PPi and Pi. S-adenosylmethionine is the source of methyl groups for most biological methylations. The encoded protein is found as a homotetramer or a homodimer whereas a third form, MAT II , is encoded by the MAT2A gene. Mutations in this gene are associated with methionine adenosyltransferase deficiency.
Specifications
MATN1 | |
Polyclonal | |
Unconjugated | |
MATN1 | |
cartilage matrix protein; Cmp; CRTM; MAT; Mat1; MAT1A; MATA1; MATN1; matrilin 1, cartilage matrix protein; matrilin 1, cartilage matrix protein 1; matrilin-1; SAMS; SAMS1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4146 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P21941 | |
MATN1 | |
Recombinant protein corresponding to Human MATN1. Recombinant protein control fragment (Product #RP-94369). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction