Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MATN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520724
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520724 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520724 Supplier Invitrogen™ Supplier No. PA520724
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is rat liver tissue lysate. PA5-20724 can be used with blocking peptide PEP-0838. Antibodies should not be exposed to prolonged high temperatures.

This gene catalyzes a two-step reaction that involves the transfer of the adenosyl moiety of ATP to methionine to form S-adenosylmethionine and tripolyphosphate, which is subsequently cleaved to PPi and Pi. S-adenosylmethionine is the source of methyl groups for most biological methylations. The encoded protein is found as a homotetramer or a homodimer whereas a third form, MAT II , is encoded by the MAT2A gene. Mutations in this gene are associated with methionine adenosyltransferase deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MATN1
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene MATN1
Gene Accession No. P21941, P51942
Gene Alias cartilage matrix protein; Cmp; CRTM; MAT; Mat1; MAT1A; MATA1; MATN1; matrilin 1, cartilage matrix protein; matrilin 1, cartilage matrix protein 1; matrilin-1; SAMS; SAMS1
Gene Symbols MATN1
Host Species Rabbit
Immunogen A 13 amino acid peptide from near the carboxy terminus of human MATN1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17180, 297894, 4146
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.