Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MATN3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145085
Description
Immunogen sequence: ICVNDRSGSY HCECYEGYTL NEDRKTCSAQ DKCALGTHGC QHICVNDRTG SHHCECYEGY TLNADKKTCS VRDKCALGSH GC Antigen sequence identity: Mouse 84%; Rat 91%.
Matrilins (MATNs) are a family of non-collagenous extra-cellular matrix (ECM) proteins consisting of four known members that have been proposed to play key roles in modulating cellular phenotypes during chondrogenesis of mesenchymal stem cells (MSCs). MATN1 and MATN3 are expressed specifically in cartilage and are among the most up-regulated ECM proteins during chondrogenesis. MATN3 is composed of a single N-terminal von Willebrand Factor A (vWFA) domain followed by four epidermal growth factor (EGF) repeats and a coiled-coil domain whereas MATN1 is composed of two vWFA domains separated by one EGF-like domain. MATN1 or MATN3 may play a role in modulating chondrogenesis during the chondrocyte differentiation process. Mutations of this gene have been associated with variety of inherited chondrodysplasias. Recent studies show that aberrant expression and processing of MATN3 are hallmarks of conventional cartilaginous neoplasms.
Specifications
MATN3 | |
Polyclonal | |
Unconjugated | |
MATN3 | |
AV009181; B14.5b; CI-B14.5b; DIPOA; EDM5; HOA; Matn3; matrilin 3; matrilin-3; NADHDH2; NDUFC2; OADIP; OS2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
4148 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15232 | |
MATN3 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 320-401. Recombinant protein control fragment (Product #RP-109824). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction