Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MATN4 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA520735

Catalog No. PIPA520735


Only null left
Add to Cart

Description

Description

A suggested positive control is rat brain tissue lysate. PA5-20735 can be used with blocking peptide PEP-0849.

Matrilin (MATNs) are a family of non-collagenous extracellular matrix proteins consisting of four known members that have been proposed to play key roles in the formation of both collagen-dependent and collagen-independent filamentous networks. Members of the matrilin family all share a structure made up of von Willebrand factor A domains, epidermal growth factor-like domains and a coiled coil alpha-helical module. MATN1 and MATN3 are expressed mainly in cartilage, while MATN2 and MATN4 occur in a wide variety of extracellular matrices such as embryonic kidney, lung, bone, cartilage and nervous system. The matrilin genes are strictly and differently regulated and their expression may serve as markers for cellular differentiation. MATN4 could serve as an odontoblast differentiation marker, e.g. in odontoblast stem cell research.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MATN4
Polyclonal
Unconjugated
MATN4
HE6WCR54; MAT-4; MATN4; matrilin 4; matrilin-4
Rabbit
Antigen affinity chromatography
RUO
17183, 296358, 8785
Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Liquid
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
O89029, O95460
MATN4
A 13 amino acid peptide near the amino terminus of human MATN4.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.