Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MATN4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596206
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596206 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596206 Supplier Invitrogen™ Supplier No. PA596206
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FGNHSCQHEC VSTPGGPRCH CREGHDLQPD GRSCQVRDLC NGVDHGCEFQ CVSEGLSYRC LCPEGRQLQA DGKSCNRCRE GHVDLVLLVD GSKSVRPQNF ELVKRFVNQI VDFLDVSPEG TRVGLVQFSS RVRTEFPLGR YGTAAEVKQA VLAVEYMERG TMTGLALRHM VEHSFSEAQG ARPRALNVPR VGLVFTDGRS QDDISVWAAR AKEEGIVMYA VGVGKAVEAE LREIASEPAE LHVSYAPDFG TMTHLLENLR GSICPEEGIS AGTELRSPCE CESLVEFQGR TLGALESLTL NLAQLTARLE DLENQLANQK; Positive Samples: HepG2, NCI-H460, Mouse heart, Rat kidney.

Matrilin (MATNs) are a family of non-collagenous extracellular matrix proteins consisting of four known members that have been proposed to play key roles in the formation of both collagen-dependent and collagen-independent filamentous networks. Members of the matrilin family all share a structure made up of von Willebrand factor A domains, epidermal growth factor-like domains and a coiled coil alpha-helical module. MATN1 and MATN3 are expressed mainly in cartilage, while MATN2 and MATN4 occur in a wide variety of extracellular matrices such as embryonic kidney, lung, bone, cartilage and nervous system. The matrilin genes are strictly and differently regulated and their expression may serve as markers for cellular differentiation. MATN4 could serve as an odontoblast differentiation marker, e.g. in odontoblast stem cell research.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MATN4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MATN4
Gene Accession No. O89029, O95460
Gene Alias HE6WCR54; MAT-4; MATN4; matrilin 4; matrilin-4
Gene Symbols MATN4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 221-540 of human MATN4 (NP_0850801).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17183, 296358, 8785
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.