Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MATR3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557719
Description
Immunogen sequence: VVHIMDFQRG KNLRYQLLQL VEPFGVISNH LILNKINEAF IEMATTEDAQ AAVDYYTTTP ALVFGKPVRV HLSQKYKRIK KPEGKPDQKF Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
This gene encodes a nuclear matrix protein, which is proposed to stabilize certain messenger RNA species. Mutations of this gene are associated with distal myopathy 2, which often includes vocal cord and pharyngeal weakness. Alternatively spliced transcript variants, including read-through transcripts composed of the upstream small nucleolar RNA host gene 4 (non-protein coding) and matrin 3 gene sequence, have been identified. Pseudogenes of this gene are located on chromosomes 1 and X.
Specifications
| MATR3 | |
| Polyclonal | |
| Unconjugated | |
| MATR3 | |
| 1110061A14Rik; 2810017I02Rik; AI841759; ALS21; AW555618; D030046F20Rik; KIAA0723; MATR3; matrin 3; matrin-3; matrin-3-like protein; mKIAA0723; MPD2; nuclear scaffold protein P130/MAT3; P130/MAT3; VCPDM; vocal cord and pharyngeal weakness with distal myopathy | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9782 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P43243 | |
| MATR3 | |
| Recombinant protein corresponding to Human MATR3. Recombinant protein control fragment (Product #RP-96788). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction