Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MATR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557720
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557720 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557720 Supplier Invitrogen™ Supplier No. PA557720
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LKRRRTEEGP TLSYGRDGRS ATREPPYRVP RDDWEEKRHF RRDSFDDRGP SLNPVLDYDH GSRSQESGYY DRMDYEDDRL RDGERCRDD Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene encodes a nuclear matrix protein, which is proposed to stabilize certain messenger RNA species. Mutations of this gene are associated with distal myopathy 2, which often includes vocal cord and pharyngeal weakness. Alternatively spliced transcript variants, including read-through transcripts composed of the upstream small nucleolar RNA host gene 4 (non-protein coding) and matrin 3 gene sequence, have been identified. Pseudogenes of this gene are located on chromosomes 1 and X.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MATR3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MATR3
Gene Accession No. P43243, Q8K310
Gene Alias 1110061A14Rik; 2810017I02Rik; AI841759; ALS21; AW555618; D030046F20Rik; KIAA0723; MATR3; matrin 3; matrin-3; matrin-3-like protein; mKIAA0723; MPD2; nuclear scaffold protein P130/MAT3; P130/MAT3; VCPDM; vocal cord and pharyngeal weakness with distal myopathy
Gene Symbols MATR3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MATR3. Recombinant protein control fragment (Product #RP-96789).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17184, 9782
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.