Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Matrin 3 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1001559AF350
Description
Matrin 3 Polyclonal antibody specifically detects Matrin 3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Matrin 3 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
distal 2, DKFZp686K0542, DKFZp686K23100, matrin 3, matrin-3, vocal cord and pharyngeal weakness with distal myopathy | |
The immunogen recognized by this antibody maps to a region between residues 475 and 500 of human Matrin 3 using the numbering given in entry NP_061322.2 (GeneID 9782). | |
0.1 mL | |
Cancer | |
9782 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction