Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrine 98.0+%, TCI America™
SDP

Catalog No. M21201G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21201G 1g
M2120200MG 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21201G Supplier TCI America Supplier No. M21201G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 519-02-8
Molecular Formula C15H24N2O

Specifications

Quantity 1g
Melting Point 85°C
Boiling Point 223°C
Formula Weight 248.37
Physical Form Crystalline Powder at 20°C
Color White
Purity ≥98.0% (GC,T)
Chemical Name or Material Matrine
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.