Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matriptase 2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988442
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8442 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8442 Supplier Bioss Supplier No. BS8296R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMPRSS6 is a serine protease which hydrolyzes a range of proteins including type I collagen, fibronectin and fibrinogen. TMPRSS6 can also activate urokinase-type plasminogen activator with low efficiency. TMPRSS6 may play a specialized role in matrix remodeling processes in liver. TMPRSS6 is required to sense iron deficiency. Overexpression of TMPRSS6 suppresses activation of the HAMP promoter. The protein encoded by this gene is a type II transmembrane serine proteinase that is found attached to the cell surface. The encoded protein may be involved in matrix remodeling processes in the liver.

Specifications

Antigen Matriptase 2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TMPRSS6
Gene Accession No. Q8IU80
Gene Alias 1300008A22Rik; IRIDA; matriptase 2; Matriptase-2; membrane-bound mosaic serine proteinase matriptase-2; TMPRSS6; Transmembrane protease serine 6; transmembrane protease, serine 6; transmembrane serine protease 6; type II transmembrane serine protease 6; UNQ354/PRO618
Gene Symbols TMPRSS6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Matriptase 2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 164656
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.