Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Matriptase/ST14 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002308G
Description
Matriptase/ST14 Polyclonal antibody specifically detects Matriptase/ST14 in Human samples. It is validated for Western BlotSpecifications
Matriptase/ST14 | |
Polyclonal | |
Western Blot | |
EC 3.4.21, HAI, Matriptase, Membrane-type serine protease 1, MT-SP1EC 3.4.21.109, prostamin, PRSS14, Serine protease 14, Serine protease TADG-15, SNC19MTSP1, suppression of tumorigenicity 14 (colon carcinoma), suppression of tumorigenicity 14 (colon carcinoma, matriptase, epithin), suppressor of tumorigenicity 14 protein, TADG15, TMPRSS14, tumor associated differentially expressed gene 15 protein, Tumor-associated differentially-expressed gene 15 protein | |
The immunogen recognized by this antibody maps to a region between residues 800 and the C-terminus (residue 855) of human Membrane-type serine protease 1 using the numbering given in SwissProt entry Q9Y5Y6 (GeneID 6768). | |
0.1 mL | |
Cancer | |
6768 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction