Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Matrix™ Filtered Pipette Tips
Click to view available options
Volume (Metric):
12.5 μL
1250 μL
For Use With (Equipment):
Thermo Scientific Matrix
Thermo Scientific Matrix Impact
Description
Eliminate pipetter barrel contamination with Thermo Scientific™ Matrix™ Filtered Pipette Tips. Rigorously tested to ensure optimum performance, these filtered tips eliminate the risk of cross-contamination from aerosols created while pipetting genomic material, bacterial samples or other precious samples.
Benefits:
- Stringent testing guarantees filter tips that effectively block aerosols.
- Hydrophobic polyethylene filter material ensures no particulates contaminate samples.
- Inadvertent contact with filter will not contaminate sample or result in sample loss.
- Easy to open packaging maintains product sterility while providing easy access.
- Hinged lid allows rack to be opened without laying lid on benchtop, helping maintain sterility.
- Racks are sealed in thick polyethylene bag for sterility, which is easy to open with simple snap-and-tear packaging.
Stringent Filter Testing:
- Filter must meet certain porosity requirements and must form a complete barrier across face of tip.
- To ensure defects are not present, two specific tests are performed: mercury intrusion and particle count testing.
- Mercury Intrusion Testing (QC 13.1 - QC 13.8) ensures the filter itself meets specific aerosol blocking requirements.
- Particle Count Testing (Test Protocol QAS-0100) ensures filter is effective at blocking aerosols after installation into tip; also ensures proper shape and fit of filter.
- Every lot of tips is tested by drawing a vacuum of aerosols against the filter and ensuring particles do not pass through.
Recommended for:
- Bacteriology
- Gel loading
- PCR setup
- Preventing cross-contamination during routine pipetting

Specifications
Specifications
Volume (Metric) | 1250 μL |
Filtered | Filtered |
Sterility | Sterile |
Format | Rack |
For Use With (Equipment) | Thermo Scientific Matrix Impact |
Product Line | Matrix |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction