Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Matrix Modifier, 0.2% Pd (2000 ppm Pd), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
50 mL
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
Specifications
Specifications
Chemical Name or Material | Matrix Modifier |
Packaging | Natural Poly Bottle |
Concentration or Composition Notes | 0.2% Pd,2000ppm Pd |
CAS Min % | 92.0, 6.16, 0.15, 0.03 |
CAS Max % | 95.0, 6.42, 0.15, 0.03 |
CAS | 7697-37-2, 7440-05-3, 7647-01-0 |
Physical Form | Liquid |
Color | Brown |
Melting Point | Approximately 0°C |
Quantity | 50 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction